missing translation for 'onlineSavingsMsg'
Learn More

Mefenamic acid, 98%

CAS: 61-68-7 | C15H15NO2 | 241.29 g/mol

Supplier:  Thermo Scientific Chemicals J6270522

 View more versions of this product

Catalog No. AAJ6270522


Only null left
Explore available promotions

Description

Description

Mefenamic acid has demonstrated antiproliferative activity against colorectal cancer cells. It has Analgesic, Anti-inflammatory activity. It is used to relieve mild to moderate pain, including headaches, dental pain, osteoarthritis and rheumatoid arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Mefenamic acid has demonstrated antiproliferative activity against colorectal cancer cells. It has Analgesic, Anti-inflammatory activity. It is used to relieve mild to moderate pain, including headaches, dental pain, osteoarthritis and rheumatoid arthritis.

Solubility
It is soluble in acetone, chloroform, dichloromethane, methanol. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Keep away from strong oxidizing agents.
Specifications

Specifications

Mefenamic acid
White
MFCD00051721
14,5798
It is soluble in acetone,chloroform,dichloromethane,methanol. Insoluble in water.
CC1=C(C(=CC=C1)NC2=CC=CC=C2C(=O)O)C
241.29
CHEBI:6717
98%
61-68-7
C15H15NO2
100 g
mefenamic acid, ponstel, ponstan, 2-2,3-dimethylphenyl amino benzoic acid, mephenamic acid, parkemed, coslan, mefacit, ponalar, methenamic acid
HYYBABOKPJLUIN-UHFFFAOYSA-N
2-(2,3-dimethylanilino)benzoic acid
4044
241.29
Powder

Chemical Identifiers

61-68-7
241.29
HYYBABOKPJLUIN-UHFFFAOYSA-N
4044
2-(2,3-dimethylanilino)benzoic acid
C15H15NO2
MFCD00051721
mefenamic acid, ponstel, ponstan, 2-2,3-dimethylphenyl amino benzoic acid, mephenamic acid, parkemed, coslan, mefacit, ponalar, methenamic acid
CHEBI:6717
CC1=C(C(=CC=C1)NC2=CC=CC=C2C(=O)O)C
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

P264b-P270-P301+P312-P330-P501c

H302

EINECSNumber : 200-513-1

RTECSNumber : CB4550000

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only