missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mecobalamin, JP, 98-101%, Spectrum™ Chemical
M1804, 13422-55-4, C63H91CoN13O14P
$1308.66 - $99528.37
Chemical Identifiers
| CAS | 13422-55-4 |
|---|---|
| Molecular Formula | C63H91CoN13O14P |
| Molecular Weight (g/mol) | 1344.41 |
| InChI Key | QXOMMGINPOYKPR-UHFFFAOYNA-L |
| IUPAC Name | λ²-cobalt(2+) 8,13,18-tris(2-carbamoylethyl)-3,14,19-tris(carbamoylmethyl)-4-{2-[(2-{[5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl phosphonato]oxy}propyl)carbamoyl]ethyl}-1,4,6,9,9,14,16,19-octamethyl-20,21,22,23-tetraazapentacyclo[15.2.1.1²,âµ.1â·,¹â°.1¹²,¹âµ]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-ide methanide |
| SMILES | [CH3-].[Co++].CC(CNC(=O)CCC1(C)C(CC(N)=O)C2N=C1C(C)=C1N=C(C=C3N=C(C(C)=C4[N-]C2(C)C(C)(CC(N)=O)C4CCC(N)=O)C(C)(CC(N)=O)C3CCC(N)=O)C(C)(C)C1CCC(N)=O)OP([O-])(=O)OC1C(CO)OC(C1O)N1C=NC2=CC(C)=C(C)C=C12 |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18604257
|
Spectrum Chemical Mfg Cor
M1804500MG |
500 mg | Amber Glass Bottle |
Each for $1,308.66
|
|
||||
|
18604258
|
Spectrum Chemical Mfg Cor
M18041GM |
1 g | Amber Glass Bottle |
Each for $2,240.23
|
|
||||
|
18604259
|
Spectrum Chemical Mfg Cor
M18045GM |
5 g | Amber Glass Bottle |
Each for $8,462.94
|
|
||||
|
18604260
|
Spectrum Chemical Mfg Cor
M180410GM |
10 g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18604261
|
Spectrum Chemical Mfg Cor
M180425GM |
25 g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18604262
|
Spectrum Chemical Mfg Cor
M1804100GMBL |
100 g | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
|
18604263
|
Spectrum Chemical Mfg Cor
M18041KGBL |
1 kg | Amber Glass Bottle |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Description
Spectrum™ Chemical Mecobalamin, JP is equivalent physiologically to vitamin B12, and can be used to prevent or treat pathology arising from vitamin B12 deficiency. All Spectrum Chemical JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Chemical Identifiers
| 13422-55-4 | |
| 1344.41 | |
| λ²-cobalt(2+) 8,13,18-tris(2-carbamoylethyl)-3,14,19-tris(carbamoylmethyl)-4-{2-[(2-{[5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl phosphonato]oxy}propyl)carbamoyl]ethyl}-1,4,6,9,9,14,16,19-octamethyl-20,21,22,23-tetraazapentacyclo[15.2.1.1²,âµ.1â·,¹â°.1¹²,¹âµ]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-ide methanide |
| C63H91CoN13O14P | |
| QXOMMGINPOYKPR-UHFFFAOYNA-L | |
| [CH3-].[Co++].CC(CNC(=O)CCC1(C)C(CC(N)=O)C2N=C1C(C)=C1N=C(C=C3N=C(C(C)=C4[N-]C2(C)C(C)(CC(N)=O)C4CCC(N)=O)C(C)(CC(N)=O)C3CCC(N)=O)C(C)(C)C1CCC(N)=O)OP([O-])(=O)OC1C(CO)OC(C1O)N1C=NC2=CC(C)=C(C)C=C12 |
Specifications
| 13422-55-4 | |
| Amber Glass Bottle | |
| 500 mg | |
| [CH3-].[Co++].CC(CNC(=O)CCC1(C)C(CC(N)=O)C2N=C1C(C)=C1N=C(C=C3N=C(C(C)=C4[N-]C2(C)C(C)(CC(N)=O)C4CCC(N)=O)C(C)(CC(N)=O)C3CCC(N)=O)C(C)(C)C1CCC(N)=O)OP([O-])(=O)OC1C(CO)OC(C1O)N1C=NC2=CC(C)=C(C)C=C12 | |
| 1344.41 | |
| JP |
| 100% | |
| C63H91CoN13O14P | |
| QXOMMGINPOYKPR-UHFFFAOYNA-L | |
| λ²-cobalt(2+) 8,13,18-tris(2-carbamoylethyl)-3,14,19-tris(carbamoylmethyl)-4-{2-[(2-{[5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl phosphonato]oxy}propyl)carbamoyl]ethyl}-1,4,6,9,9,14,16,19-octamethyl-20,21,22,23-tetraazapentacyclo[15.2.1.1²,âµ.1â·,¹â°.1¹²,¹âµ]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-ide methanide | |
| 98 to 101% |