missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mecobalamin, JP, 98-101%, Spectrum™ Chemical
M1804, 13422-55-4, C63H91CoN13O14P
Supplier: Spectrum Chemical Mfg Cor M180410GM
Description
Spectrum™ Chemical Mecobalamin, JP is equivalent physiologically to vitamin B12, and can be used to prevent or treat pathology arising from vitamin B12 deficiency. All Spectrum Chemical JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 13422-55-4 | |
| Amber Glass Bottle | |
| 10 g | |
| [CH3-].[Co++].CC(CNC(=O)CCC1(C)C(CC(N)=O)C2N=C1C(C)=C1N=C(C=C3N=C(C(C)=C4[N-]C2(C)C(C)(CC(N)=O)C4CCC(N)=O)C(C)(CC(N)=O)C3CCC(N)=O)C(C)(C)C1CCC(N)=O)OP([O-])(=O)OC1C(CO)OC(C1O)N1C=NC2=CC(C)=C(C)C=C12 | |
| 1344.41 | |
| JP |
| 100% | |
| C63H91CoN13O14P | |
| QXOMMGINPOYKPR-UHFFFAOYNA-L | |
| λ²-cobalt(2+) 8,13,18-tris(2-carbamoylethyl)-3,14,19-tris(carbamoylmethyl)-4-{2-[(2-{[5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl phosphonato]oxy}propyl)carbamoyl]ethyl}-1,4,6,9,9,14,16,19-octamethyl-20,21,22,23-tetraazapentacyclo[15.2.1.1²,âµ.1â·,¹â°.1¹²,¹âµ]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-ide methanide | |
| 98 to 101% |
Chemical Identifiers
| 13422-55-4 | |
| 1344.41 | |
| λ²-cobalt(2+) 8,13,18-tris(2-carbamoylethyl)-3,14,19-tris(carbamoylmethyl)-4-{2-[(2-{[5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl phosphonato]oxy}propyl)carbamoyl]ethyl}-1,4,6,9,9,14,16,19-octamethyl-20,21,22,23-tetraazapentacyclo[15.2.1.1²,âµ.1â·,¹â°.1¹²,¹âµ]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-ide methanide |
| C63H91CoN13O14P | |
| QXOMMGINPOYKPR-UHFFFAOYNA-L | |
| [CH3-].[Co++].CC(CNC(=O)CCC1(C)C(CC(N)=O)C2N=C1C(C)=C1N=C(C=C3N=C(C(C)=C4[N-]C2(C)C(C)(CC(N)=O)C4CCC(N)=O)C(C)(CC(N)=O)C3CCC(N)=O)C(C)(C)C1CCC(N)=O)OP([O-])(=O)OC1C(CO)OC(C1O)N1C=NC2=CC(C)=C(C)C=C12 |