missing translation for 'onlineSavingsMsg'
Learn More

Mecobalamin, JP, 98-101%, Spectrum™ Chemical

Catalog No. 18604263
Change view
Click to view available options
Quantity:
500 mg
1 g
5 g
10 g
25 g
100 g
1 kg
Packaging:
Amber Glass Bottle
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604263 1 kg Amber Glass Bottle
18604257 500 mg Amber Glass Bottle
18604258 1 g Amber Glass Bottle
18604259 5 g Amber Glass Bottle
18604260 10 g Amber Glass Bottle
18604261 25 g Amber Glass Bottle
18604262 100 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 18604263 Supplier Spectrum Chemical Mfg Cor Supplier No. M18041KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1804, 13422-55-4, C63H91CoN13O14P

Spectrum™ Chemical Mecobalamin, JP is equivalent physiologically to vitamin B12, and can be used to prevent or treat pathology arising from vitamin B12 deficiency. All Spectrum Chemical JP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 13422-55-4
Molecular Formula C63H91CoN13O14P
Molecular Weight (g/mol) 1344.41
InChI Key QXOMMGINPOYKPR-UHFFFAOYNA-L
IUPAC Name λ²-cobalt(2+) 8,13,18-tris(2-carbamoylethyl)-3,14,19-tris(carbamoylmethyl)-4-{2-[(2-{[5-(5,6-dimethyl-1H-1,3-benzodiazol-1-yl)-4-hydroxy-2-(hydroxymethyl)oxolan-3-yl phosphonato]oxy}propyl)carbamoyl]ethyl}-1,4,6,9,9,14,16,19-octamethyl-20,21,22,23-tetraazapentacyclo[15.2.1.1²,⁵.1⁷,¹⁰.1¹²,¹⁵]tricosa-5(23),6,10(22),11,15(21),16-hexaen-20-ide methanide
SMILES [CH3-].[Co++].CC(CNC(=O)CCC1(C)C(CC(N)=O)C2N=C1C(C)=C1N=C(C=C3N=C(C(C)=C4[N-]C2(C)C(C)(CC(N)=O)C4CCC(N)=O)C(C)(CC(N)=O)C3CCC(N)=O)C(C)(C)C1CCC(N)=O)OP([O-])(=O)OC1C(CO)OC(C1O)N1C=NC2=CC(C)=C(C)C=C12

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 1 kg
Percent Purity 98 to 101%
Grade JP
Chemical Name or Material Mecobalamin