missing translation for 'onlineSavingsMsg'
Learn More

Meclofenoxate Hydrochloride 98.0+%, TCI America™

Catalog No. M24601G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M24601G 1 g
M24605G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M24601G Supplier TCI America Supplier No. M24601G

Chemical Identifiers

CAS 3685-84-5
Molecular Formula C12H17Cl2NO3
Molecular Weight (g/mol) 294.17
MDL Number MFCD00012533
InChI Key FIVHOHCAXWQPGC-UHFFFAOYSA-N
Synonym meclofenoxate hydrochloride, centrophenoxine hydrochloride, lucidril, cerutil, meclofenoxate hcl, atsefen, helfergin, marucotol, acefen, brenal
PubChem CID 19379
IUPAC Name hydrogen 2-(dimethylamino)ethyl 2-(4-chlorophenoxy)acetate chloride
SMILES [H+].[Cl-].CN(C)CCOC(=O)COC1=CC=C(Cl)C=C1

Specifications

Melting Point 139°C
Color White
Quantity 1 g
Formula Weight 294.17
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Meclofenoxate Hydrochloride
RTECSNumber AG0440000
TSCA No
Recommended Storage Refrigerator