accessibility menu, dialog, popup

UserName

HClH3CNCH3OOOCl

CAS RN 3685-84-5

IUPAC Name: hydrogen 2-(dimethylamino)ethyl 2-(4-chlorophenoxy)acetate chloride
Synonyms: meclofenoxate hydrochloride
Molecular Weight (g/mol): 294.17
Molecular Formula: C12H17Cl2NO3
InChi Key: FIVHOHCAXWQPGC-UHFFFAOYSA-N
SMILES: [H+].[Cl-].CN(C)CCOC(=O)COC1=CC=C(Cl)C=C1
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (2)