missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Meclizine Dihydrochloride 98.0+%, TCI America™
Supplier: TCI America M27555G
Specifications
| Meclizine Dihydrochloride | |
| 212°C | |
| C25H29Cl3N2 | |
| 5 g | |
| VCTHNOIYJIXQLV-UHFFFAOYSA-N | |
| 1-[(4-chlorophenyl)-phenylmethyl]-4-[(3-methylphenyl)methyl]piperazine;dihydrochloride | |
| 64713 | |
| ≥98.0% (HPLC) |
| 1104-22-9 | |
| White | |
| MFCD00058199 | |
| meclizine dihydrochloride, meclozine dihydrochloride, bonamine, meclizine hydrochloride, postafen, vertizine, taizer, v-cline, 1-4-chlorophenyl phenyl methyl-4-3-methylbenzyl piperazine dihydrochloride, duremesin | |
| CC1=CC(=CC=C1)CN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=C(C=C4)Cl.Cl.Cl | |
| 463.871 | |
| 463.87 | |
| Crystalline Powder |
Chemical Identifiers
| 1104-22-9 | |
| 463.871 | |
| VCTHNOIYJIXQLV-UHFFFAOYSA-N | |
| 64713 | |
| CC1=CC(=CC=C1)CN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=C(C=C4)Cl.Cl.Cl |
| C25H29Cl3N2 | |
| MFCD00058199 | |
| meclizine dihydrochloride, meclozine dihydrochloride, bonamine, meclizine hydrochloride, postafen, vertizine, taizer, v-cline, 1-4-chlorophenyl phenyl methyl-4-3-methylbenzyl piperazine dihydrochloride, duremesin | |
| 1-[(4-chlorophenyl)-phenylmethyl]-4-[(3-methylphenyl)methyl]piperazine;dihydrochloride |
Safety and Handling
RTECSNumber : TL2005000
TSCA : Yes
Recommended Storage : Refrigerator