missing translation for 'onlineSavingsMsg'
Learn More

Meclizine Dihydrochloride 98.0+%, TCI America™

Catalog No. p-7133408
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M27555G 5 g
M275525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M27555G Supplier TCI America Supplier No. M27555G
Only null left

Chemical Identifiers

CAS 1104-22-9
Molecular Formula C25H29Cl3N2
Molecular Weight (g/mol) 463.871
MDL Number MFCD00058199
InChI Key VCTHNOIYJIXQLV-UHFFFAOYSA-N
Synonym meclizine dihydrochloride, meclozine dihydrochloride, bonamine, meclizine hydrochloride, postafen, vertizine, taizer, v-cline, 1-4-chlorophenyl phenyl methyl-4-3-methylbenzyl piperazine dihydrochloride, duremesin
PubChem CID 64713
IUPAC Name 1-[(4-chlorophenyl)-phenylmethyl]-4-[(3-methylphenyl)methyl]piperazine;dihydrochloride
SMILES CC1=CC(=CC=C1)CN2CCN(CC2)C(C3=CC=CC=C3)C4=CC=C(C=C4)Cl.Cl.Cl

Specifications

Melting Point 212°C
Color White
Quantity 5 g
Formula Weight 463.87
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Meclizine Dihydrochloride
RTECSNumber TL2005000
TSCA Yes
Recommended Storage Refrigerator