missing translation for 'onlineSavingsMsg'
Learn More

Mebendazole 98.0+%, TCI America™

Supplier:  TCI America M22735G

 View more versions of this product

Catalog No. M22735G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Mebendazole
287°C
C16H13N3O3
5 g
OPXLLQIJSORQAM-UHFFFAOYSA-N
methyl N-(6-benzoyl-1H-1,3-benzodiazol-2-yl)carbamate
4030
295.30
Crystalline Powder
31431-39-7
White-Yellow
MFCD00057872
mebendazole, vermox, telmin, mebenvet, pantelmin, vermirax, mebenoazole, ovitelmin, bantenol, mebutar
COC(=O)NC1=NC2=CC=C(C=C2N1)C(=O)C1=CC=CC=C1
295.30
CHEBI:6704
≥98.0% (HPLC,N)

Chemical Identifiers

31431-39-7
295.30
OPXLLQIJSORQAM-UHFFFAOYSA-N
4030
methyl N-(6-benzoyl-1H-1,3-benzodiazol-2-yl)carbamate
C16H13N3O3
MFCD00057872
mebendazole, vermox, telmin, mebenvet, pantelmin, vermirax, mebenoazole, ovitelmin, bantenol, mebutar
CHEBI:6704
COC(=O)NC1=NC2=CC=C(C=C2N1)C(=O)C1=CC=CC=C1
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : EY8600000

TSCA : No

SDS
Documents

Documents

Promotions

Promotions