missing translation for 'onlineSavingsMsg'
Learn More

Mebendazole 98.0+%, TCI America™

Catalog No. p-7133128
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M22735G 5 g
M227325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M22735G Supplier TCI America Supplier No. M22735G
Only null left

Chemical Identifiers

CAS 31431-39-7
Molecular Formula C16H13N3O3
Molecular Weight (g/mol) 295.30
MDL Number MFCD00057872
InChI Key OPXLLQIJSORQAM-UHFFFAOYSA-N
Synonym mebendazole, vermox, telmin, mebenvet, pantelmin, vermirax, mebenoazole, ovitelmin, bantenol, mebutar
PubChem CID 4030
ChEBI CHEBI:6704
IUPAC Name methyl N-(6-benzoyl-1H-1,3-benzodiazol-2-yl)carbamate
SMILES COC(=O)NC1=NC2=CC=C(C=C2N1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 287°C
Color White-Yellow
Quantity 5 g
Formula Weight 295.30
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Mebendazole
RTECSNumber EY8600000
TSCA No