missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ MCF7 (+hIGF-1) Control Lysate (1 mg/mL)

Catalog No. PIA37959
Change view
Click to view available options
Quantity:
0.5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIA37959 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIA37959 Supplier Thermo Scientific™ Supplier No. A37959

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Thermo Scientific™ MCF7 (+hIGF-1) Control Lysate is a stimulated cell lysate that has been validated for the expression levels and phosphorylation of key targets in the AKT signaling pathway.

  • Validated for the detection and phosphorylation of AKT, PTEN, IRS1, IGF1-R, GSK3a, GSK3b, PRAS40, mTOR, p70S6K, and TSC2 proteins
  • Optimized - stimulation and lysis conditions optimized for detection of AKT signaling targets
  • Convenient - lysate is ready-to-use for immunoprecipitation, immunoprecipitation to mass spectrometry, and western blotting applications
  • Concentration: 1 mg/mL
  • Stable for two years at -80°C

Specifications

Detection Method Mass Spectrometry
For Use With (Equipment) Mass Spectrometer
Shipping Condition Dry Ice
Workflow Step Sample Enrichment
Form Frozen Liquid
Product Line Thermo Scientific
Quantity 0.5 mL
Type Control Lysate
Final Product Type Protein
Starting Material Cell Lysate
What are the characteristics of MCF7 (+hIGF-1) Control Lysate, and what is its main purpose?

MCF7 (+hIGF-1) Control Lysate is a stimulated MCF-7 cell lysate that has been validated for the expression levels and phosphorylation of key targets (AKT, PTEN, IRS1, IGF1-R, GSK3a, GSK3b, PRAS40, mTOR, p70S6K, and TSC2) in the AKT signaling pathway. It serves as a positive control for use with SureQuant AKT panel kits.