missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ McCoy's 5A (Modified) Medium, GlutaMAX™ Supplement

McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues.

$135.00 - $135.00

Specifications

Form Liquid
Manufacturing Quality cGMP-compliant under the ISO 13485 standard
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
36-600-021
View Documents
Gibco™
36600021
500 mL
Each for $135.00
Only null left
 
Description

Description

McCoy's 5A (modified) is a general purpose medium that supports the propagation of many types of primary cells, established cell lines, and explants from biopsy tissues. This medium will support the growth of primary mammalian cells derived from normal bone marrow, skin, spleen, kidney, lung, rat embryos, and other tissues.

This McCoy's 5A is modified as follows:

With: High Glucose, GlutaMAX™, Bacto-peptone, Phenol Red

Without: Sodium Pyruvate

The complete formulation is available.

Dr. Thomas McCoy originally formulated McCoy's 5A medium as a modification of Basal Medium 5A. Unlike other media, McCoy's 5A contains the reducing agent glutathione, bacto-peptone, and a high level of glucose. This product also includes Dr. Hsu's addition of Hanks' salts to enable use outside a CO2 incubator.

Gibco™ McCoy's 5A (modified) requires serum supplementation, commonly with 10% Fetal Bovine Serum (FBS). Gibco™ McCoy's 5A (modified) uses a sodium bicarbonate buffer system (2.2 g / L) and therefore requires a 5–10% CO2 environment to maintain physiological pH.

Specifications

Specifications

Liquid
cGMP-compliant under the ISO 13485 standard
Videos
SDS
Documents

Documents