missing translation for 'onlineSavingsMsg'
Learn More

Promega Maxwell™ RSC DNA FFPE Kit

Catalog No. PRAS1450
Encompass
Change view
Click to view available options
48 preps
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
PRAS1450 48 preps
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PRAS1450 Supplier Promega Supplier No. AS1450
Only null left

The Maxwell RSC DNA FFPE Kit is used with the Maxwell RSC Instrument to provide an easy method for efficient, automated purification of genomic DNA (gDNA) from mammalian formalin-fixed, paraffin-embedded (FFPE) tissue samples.

  • Automated DNA Extraction from FFPE Tissues
  • Extracts DNA from 1-48 formalin-fixed, paraffin-embedded (FFPE) tissue sections
  • Eliminates the use of xylene and other hazardous organic solvents
  • Purified DNA is ready to use in downstream assays including amplification assays

Specifications

Content And Storage 15°C to 30°C
No. of Reactions 48 Preps