missing translation for 'onlineSavingsMsg'
Learn More

Promega Maxwell™ 16 Purification Instruments

Catalog No. PRAS1250
Encompass
Change view
Click to view available options
Product Type:
LEV Hardware Kit
SEV Hardware Kit
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
PRAS1250 LEV Hardware Kit
PRAS1200 SEV Hardware Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PRAS1250 Supplier Promega Supplier No. AS1250
Only null left

Includes

LEV Magnetic rod assembly, LEV plunger bar adaptor, Maxwell™ 16 LEV cartridge rack, quick start guide

Requires

Either the SEV kit (Mfr. No. AS1200) or the LEV kit (Mfr. No. AS1250) to complete a functional instrument. Also requires Maxwell 16 Purification Kits.

Accessories for the Maxwell(R) 16 and Maxwell(R) 16 Forensic Instruments in either SEV or LEV Configuration

  • Accessories for the Maxwell(R) 16 and Maxwell(R) 16 Forensic Instruments in either SEV or LEV Configuration
  • Standard Elution Volume (SEV) is 200-400μL
  • Low Elution Volume (LEV) is 30-100μL
  • Plungers and Elution Tubes for the Maxwell(R) 16 Instruments only (AS2000 and AS3000)

Certifications

All Maxwell 16 instruments are labeled GPLE (General Purpose Laboratory Equipment) in the U.S.

Specifications

Content And Storage 15°C to 30°C
For Use With (Equipment) Maxwel 16 AS2000 instrument
For Use With (Application) Automated purification of genomic DNA, RNA or recombinant Proteins from a wide range of sample types
Includes Maxwell 16 instrument includes power cable and RS-232 cable for firmware upgrades. SEV Hardware Kit includes Maxwell 16 magnetic rod assembly, Maxwell 16 cartridge rack, Maxwell 16 magnetic elution rack. LEV Hardware Kit includes LEV magnetic rod assembl
Product Type LEV Hardware Kit
Quantity 1/Ea.