missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima First Strand cDNA Synthesis Kit for RT-qPCR

Catalog No. p-4526939
Change view
Click to view available options
Includes:
Kit only
Kit with dsDNase
No. of Reactions:
200 Reactions
50 Reactions
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Includes No. of Reactions
FERK1641 Kit only 50 Reactions
FERK1642 Kit only 200 Reactions
FERK1671 Kit with dsDNase 50 Reactions
FERK1672 Kit with dsDNase 200 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. FERK1641 Supplier Thermo Scientific™ Supplier No. K1641
Only null left

The Thermo Scientific Maxima First Strand cDNA Synthesis Kit for RT-qPCR, available with or without dsDNase, is a convenient system optimized for cDNA synthesis in two-step quantitative RT-PCR (RT-qPCR) applications.

The Thermo Scientific Maxima First Strand cDNA Synthesis Kit for RT-qPCR, available with or without dsDNase, is a convenient system optimized for cDNA synthesis in two-step quantitative RT-PCR (RT-qPCR) applications. It provides a dramatically simplified workflow that combines genomic DNA elimination and cDNA synthesis into one-tube procedure. These cDNA Synthesis kits use Maxima Reverse Transcriptase (RT), an advanced enzyme derived by in vitro evolution of M-MuLV RT. The enzyme features high thermostability, robustness and increased cDNA synthesis rate compared to wild type M-MuLV RT.

The Maxima First Strand cDNA Synthesis Kit for RT-qPCR is capable of reproducible cDNA synthesis from a wide range of total RNA amounts (1 pg to 5 μg) at elevated temperatures (42–65°C). The synthesis reaction can be completed in 15–30 minutes. Components of the Maxima First Strand cDNA Synthesis Kit for RT-qPCR are pre-mixed to save time and to reduce the possibility of pipetting errors.

The Maxima First Strand cDNA Synthesis Kit for RT-qPCR with dsDNase also contains a novel double-strand specific DNase engineered to remove contaminating genomic DNA from RNA preps in two minutes without damage to quality or quantity of RNA. Highly specific activity towards double-stranded DNA ensures that single-stranded DNA (such as cDNA and primers) is not cleaved and dsDNase treated RNA can be directly added to reverse transcription.

Features of the Maxima First Strand cDNA Synthesis Kit for RT-qPCR include:
• High yields of full length cDNA up to 20 kb
• Efficient cDNA synthesis at wide temperature range (42–65°C)
• Increased synthesis rate—complete cDNA synthesis in 15–30 minutes
• High sensitivity and specificity
• Integrated gDNA removal step with dsDNase kit

Maxima First Strand cDNA Synthesis Kit with dsDNase contains:
• Maxima Enzyme Mix
• 5X Reaction Mix
• nuclease-free water

Kit with dsDNase also contains
• dsDNase
• 10X dsDNase Buffer

Applications
• 2-step RT-PCR
• 2-step RT-qPCR

Additional information about reaction components
The Maxima Enzyme Mix contains Maxima Reverse Transcriptase and RiboLock RNase Inhibitor. The recombinant RiboLock RNase Inhibitor effectively protects RNA templates from degradation by RNases A, B, and C at temperatures up to 55°C.

5X Reaction Mix contains the remaining reaction components: reaction buffer, dNTPs, oligo(dT)18 and random hexamer primers. Nuclease-free water is provided for reaction set-up and dilution of sample RNA.

The absence of endo- and exo-deoxyribonucleases, ribonucleases, and phosphatases has been confirmed by appropriate quality tests.

Specifications

Content And Storage

• Maxima Enzyme Mix
• 5X Reaction Mix
• Nuclease-free water

Store at –20°C.

Format Kit
GC-Rich PCR Performance High
Reaction Speed 30 min.
Technique Reverse Transcription
Optimal Reaction Temperature 50°C to 55°C
Quantity Each
Reverse Transcriptase Maxima
Ribonuclease H Activity Yes
Shipping Condition Dry Ice
For Use With (Application) Real Time PCR (qPCR)
Final Product Type First-Strand cDNA
No. of Reactions 50 Reactions
Reaction Format Separate components
Reagent Type Reverse Transcription
Size (Final Product) Up to 20 kb
Starting Material RNA
Includes Kit only
Show More Show Less
When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H-minus RT or Maxima H-minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H-minus RTs for template-independent addition of C nucleotides.

Do Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H-minus RT, Maxima RT, and Maxima H-minus RT) possess terminal deoxynucleotidyl (TdT) activity?

All Thermo Scientific reverse transcriptases possess intrinsic TdT activity although at varying degrees depending upon the reaction conditions. For addition of template-independent C nucleotides (as for SMART and RACE experiments), this specific TdT activity can be induced by Mn2+. We would recommend Maxima H- or RevertAid H- minus RTs for this purpose.

Is it preferable to use elevated temperature in the RT reaction when using Maxima reverse transcriptases?

cDNA synthesis at higher temperatures ensures successful transcription of RNA with high levels of secondary structure, reducing issues of primer access to template. Therefore, we do recommend to use RT enzymes with high thermostability, e.g. Maxima and Maxima H Minus Reverse Transcriptases, which provide higher yields of full-length cDNA, better sensitivity, and successful transcription of GC-rich templates.

What steps should I take while performing first strand cDNA synthesis using low purity template (e.g., inhibitors in RNA sample)?

Trace amounts of reagents used in RNA purification protocols may remain in solution and inhibit first-strand synthesis, e.g., SDS, EDTA, guanidine salts, phosphate, pyrophosphate, polyamines, spermidine. To remove trace contaminants, we recommend re-precipitating the RNA with ethanol and washing the pellet with 75% ethanol, or re-purifying the RNA.

For reverse transcription, how important is the quality of RNA template?

RNA purity and integrity are essential for synthesis and quantification of cDNA. Always assess the integrity of RNA prior to cDNA synthesis. Use freshly prepared RNA. Multiple freeze/thaw cycles of the RNA sample and synthesized cDNA is not recommended. Avoid RNase contamination and discard low quality RNA.

When should I choose regular RevertAid RT or Maxima RT vs. RevertAid H Minus RT or Maxima H Minus RT?

It is generally beneficial to minimize RNase H activity when aiming to produce long transcripts for cDNA cloning. RNase H degrades RNA from RNA-DNA duplexes, which can result in truncated cDNA during reverse transcription of long mRNA. We also recommended using RNase H Minus RTs for template-independent addition of C nucleotides. In contrast, reverse transcriptases with intrinsic RNase H activity are often favored in 1-step RT-qPCR applications.

What is the fidelity of RevertAid and Maxima reverse transcriptases?

The fidelity of RevertAid and Maxima reverse transcriptases is the same as that of wild-type M-MuLV RT, which is in the range of 1 error per 15,000-27,000 nucleotides synthesized.

Do Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess terminal deoxynucleotidyl (TdT) activity?

All Thermo Scientific reverse transcriptases (RevertAid RT, RevertAid H Minus RT, Maxima RT, and Maxima H Minus RT) possess intrinsic TdT activity, although at varying degrees depending upon the reaction conditions. We recommend specialized SuperScript IV Template Switching RT Master Mix for high efficiency in applications requiring template switching RT.


For Research Use Only. Not for use in diagnostic procedures.