missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Pierceable Mat Covers for Deep Well Plates

Provides a reliable seal for every well

$1,693.03 - $1,859.47

Specifications

Color Natural
Solvent Or Alcohol Resistant No Resistance
Material Silicone
Autoclavable Autoclavable
No. of Wells 96
View More Specs

Products 2
Catalog Number Mfr. No. Sterility Price Quantity & Availability  
Catalog Number Mfr. No. Sterility Price Quantity & Availability  
14380944
View Documents Promotion Details
BRANDTECH™ BRAND™
M2200
Non-sterile
Pack of 50 Each for $1,693.03
Only null left
 
14380945
View Documents Promotion Details
BRANDTECH™ BRAND™
M2250
Sterile
Pack of 50 Each for $1,859.47
Only null left
 
Description

Description

  • Ideal for short-term storage
  • Protects samples and reduces evaporation
  • Made of pierceable silicone
  • Autoclavable
  • Available in sterile or non-sterile
  • Sealing mats will reduce the working volume of each well
  • For use with 96-deep well plates
Specifications

Specifications

Natural
Silicone
96
ANSI/SLAS
Deepwell Plate Mat
No
-80°C to +120°C
DW2200, DW2250
No Resistance
Autoclavable
50
no
None
Yes
Square
Videos
SDS
Documents

Documents