missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Pierceable Mat Covers for Deep Well Plates

Catalog No. 14380944
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
14380944 Non-sterile
14380945 Sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14380944 Supplier BRANDTECH™ BRAND™ Supplier No. M2200

Provides a reliable seal for every well

  • Ideal for short-term storage
  • Protects samples and reduces evaporation
  • Made of pierceable silicone
  • Autoclavable
  • Available in sterile or non-sterile
  • Sealing mats will reduce the working volume of each well
  • For use with 96-deep well plates

Specifications

Color Natural
Solvent Or Alcohol Resistant No Resistance
Description Sealing Mat cover for BrandTech deepwell plate DW2200. Pierceable. Pack of 50
Material Silicone
Sterility Non-sterile
Autoclavable Autoclavable
No. of Wells 96
No. per Pack 50
Format ANSI/SLAS
Pre-slit no
Product Type Deepwell Plate Mat
Adhesive None
Certified RNase/DNase-free No
Pierceable Yes
Temperature Range (Metric) -80°C to +120°C
Well Shape Square
Plate Compatibility DW2200, DW2250
Show More Show Less