missing translation for 'onlineSavingsMsg'
Learn More

BRANDTECH™ BRAND™ Pierceable Mat Covers for Deep Well Plates

Catalog No. 14380945
Change view
Click to view available options
Sterility:
Non-sterile
Sterile
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Sterility
14380945 Sterile
14380944 Non-sterile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 14380945 Supplier BRANDTECH™ BRAND™ Supplier No. M2250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Provides a reliable seal for every well

  • Ideal for short-term storage
  • Protects samples and reduces evaporation
  • Made of pierceable silicone
  • Autoclavable
  • Available in sterile or non-sterile
  • Sealing mats will reduce the working volume of each well
  • For use with 96-deep well plates

Specifications

Color Natural
Solvent Or Alcohol Resistant No Resistance
Description Plate sealing mat for BrandTech DW2250. Pierceable. Sterile, pack of 50.
Material Silicone
Sterility Sterile
Autoclavable Autoclavable
No. of Wells 96
No. per Pack 50
Format ANSI/SLAS
For Use With (Equipment) DW2200, DW2250
Pre-slit no
Product Type Deepwell Plate Mat
Adhesive None
Certified RNase/DNase-free No
Pierceable Yes
Temperature Range (Metric) -80°C to +120°C
Well Shape Square
Plate Compatibility DW2200, DW2250
Show More Show Less