missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Matrix™ ScrewTop Tube Caps, Cap Trays and Cap Inserts

Simplify sample storage with these color-coded caps and inserts, available racked in trays or in bulk to suit a variety of applications.
Supplier: Thermo Scientific™ 4470PUR
Description
Simplify sample storage with these color-coded caps and inserts. Thermo Scientific™ Matrix™ ScrewTop Tube caps and inserts are available racked in trays or in bulk to suit a variety of applications.
Superior Cap Design
- Gasketed caps ensure critical sample protection over the storage lifetime
- Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
- Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
- ScrewTop caps are compatible with storage down to -196ºC
Cap Options Simplify Sample Identification
- Cap trays feature standard microplate format compatible with automated equipment
- Caps are available in seven color options to enable sample identification
- Color inserts snap into cap for a secure fit; inserts are removable
- Inserts are available in six colors
All caps and inserts are supplied free from DNA, RNase, DNase, endotoxins and cytotoxins
Chemical Identifiers
| Sterile and PCR Ready |
Specifications
| Screw Cap | |
| Polypropylene, Silicone | |
| Bulk, 500 Caps | |
| Non-proprietary, 12x12 Data-matrix with ECC200 Built-in Error Correction | |
| 500 μL and 1.0 mL tubes | |
| Sterile and PCR Ready |
| Not Autoclavable | |
| Purple | |
| Matrix | |
| Sterile | |
| ScrewTop Tube |