missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Matrix oil, 75 centistoke hydrocarbon oil
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Odor | Odorless |
| Flash Point | >110°C (230°F) |
| Physical Form | Oily Liquid |
| Color | Colorless |
| Quantity | 1 pt. |
| Chemical Name or Material | Matrix oil |
Safety and Handling
| Hazard Category | H304 |
| Precautionary Statement | P301+P310-P331-P501c |
| EINECSNumber | 232-455-7 |
| RTECSNumber | PY8047000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only