missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Matrix - Green (Bulk Matrix from FastRNA Green)

Matrix - Green (Bulk Matrix from FastRNA Green) has 1.4 mm ceramic spheres that are ready for CE IVD labeling of clinical applications. For research use only.

Supplier:  MP Biomedicals™ 116040801

Catalog No. MP116040801


Only null left
Explore available promotions

Description

Description

  • 1.4 mm ceramic spheres
  • Ready for CE IVD labeling of clinical applications
  • For Research use only
Specifications

Specifications

Zirconium Oxide Coated Beads
0.06 in.
FastPrep™ instruments or any bead beater.
Ceramic spheres
Off White
1.4 mm
Bulk Packaged
1 kg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions