missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Matrine, TCI America™

$271.95 - $957.94

Chemical Identifiers

CAS 519-02-8
Molecular Formula C15H24N2O
Molecular Weight (g/mol) 248.37
MDL Number MFCD00210527
InChI Key ZSBXGIUJOOQZMP-JLNYLFASSA-N
Synonym Matridin-15-one, (7aS,13aR,13bR,13cS)-Dodecahydro-1H,5H,10H-dipyrido[2,1-f:3′C,2′C,1′C-ij][1,6]naphthyridin-10-one, alphaGÇÉMatrine, Sophocarpidine
PubChem CID 91466
ChEBI CHEBI:6700
SMILES C1CC2C3CCCN4C3C(CCC4)CN2C(=O)C1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
M2120200MG View Documents TCI America
M2120200MG
200mg Each for $271.95
 
M21201G View Documents TCI America
M21201G
1g Each for $957.94
 
Specifications

Chemical Identifiers

519-02-8
248.37
ZSBXGIUJOOQZMP-JLNYLFASSA-N
91466
C1CC2C3CCCN4C3C(CCC4)CN2C(=O)C1
C15H24N2O
MFCD00210527
Matridin-15-one, (7aS,13aR,13bR,13cS)-Dodecahydro-1H,5H,10H-dipyrido[2,1-f:3′C,2′C,1′C-ij][1,6]naphthyridin-10-one, alphaGÇÉMatrine, Sophocarpidine
CHEBI:6700

Specifications

223°C
85°C
519-02-8
MFCD00210527
ZSBXGIUJOOQZMP-JLNYLFASSA-N
248.37
CHEBI:6700
Crystal-Powder at 20°C
Matrine
White
200mg
C15H24N2O
Matridin-15-one, (7aS,13aR,13bR,13cS)-Dodecahydro-1H,5H,10H-dipyrido[2,1-f:3′C,2′C,1′C-ij][1,6]naphthyridin-10-one, alphaGÇÉMatrine, Sophocarpidine
C1CC2C3CCCN4C3C(CCC4)CN2C(=O)C1
91466
248.37
≥98.0% (GC,T)
Safety and Handling

Safety and Handling

RTECSNumber : OQ1754000

TSCA : No

Recommended Storage : Refrigerator