missing translation for 'onlineSavingsMsg'
Learn More

Matrine 98.0+%, TCI America™

Catalog No. M2120200MG
Change view
Click to view available options
Quantity:
200mg
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2120200MG 200mg
M21201G 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M2120200MG Supplier TCI America Supplier No. M2120200MG

Chemical Identifiers

CAS 519-02-8
Molecular Formula C15H24N2O

Specifications

Quantity 200mg
Melting Point 85°C
Boiling Point 223°C
Formula Weight 248.37
Physical Form Crystalline Powder at 20°C
Color White
Purity ≥98.0% (GC,T)
Chemical Name or Material Matrine
Recommended Storage Refrigerator