missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Matrine

$221.51 - $617.72

Chemical Identifiers

CAS 519-02-8
Molecular Formula C15H24N2O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AC466590010
View Documents
Thermo Scientific Chemicals
466590010
1 g
Each for $617.72
Only null left
 
AC466592500
View Documents
Thermo Scientific Chemicals
466592500
250 mg
Each for $221.51
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

519-02-8
C15H24N2O

Specifications

519-02-8
76.0°C
248.37
O=C1CCC[C@@H]2[C@H]3CCCN4CCC[C@@H](CN12)[C@@H]34
248.37
1 g
C15H24N2O
ZSBXGIUJOOQZMP-JLNYLFASSA-N
(1R,2R,9S,17S)-7,13-diazatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadecan-6-one
Matrine
Videos
Safety and Handling

Safety and Handling

GHS H Statement Harmful if swallowed.

Causes serious eye irritation.

GHS P Statement IF SWALLOWED: rinse mouth.

Do NOT induce vomiting.

Call a POISON CENTER or doctor/physician if you feel unwell.

Wash face, hands and any exposed skin thoroughly after handling.

Wear eye protection/face protection.

IF IN EYES: Rinse cautiously with water for several minutes.

Remove contact lenses, if present and easy to do.

Continue rinsing.

GHS Signal Word: Warning

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only