missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAST4 Monoclonal Antibody (K56010_15H12), Invitrogen™
Mouse Monoclonal Antibody
$1,461.54 - $2,194.06
Specifications
| Antigen | MAST4 |
|---|---|
| Clone | K56010_15H12 |
| Concentration | 1 mg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
Description
Sequence of this protein is as follows: KSAEPPRSPL LKRVQSEEKL SPSYGSDKKH LCSRKHSLEV TQEEVQREQS QREAPLQSLD ENVCDVPPLS RARPVEQGCL KRPVSRKVGR QESVDDLDRD KLKAKVVVKK ADGFPEKQES HQKSHGPGSD LENFALFKLE EREKKVYPKA VERSSTFENK ASMQEAPPLG SLLKDALHKQ ASVRASEGAM SDGRVPAEHR QGGGDFRRAP APGTLQDGLC HSLDRGISGK GEGTEKSSQA KELLRCEKLD SKLANIDYLR KKMSLEDKED NLCPVLKPKM TAGSHECLPG NPVRPTGGQQ EPPPASESRA
MAST4 Monoclonal antibody specifically detects MAST4 in Human samples. It is validated for Western BlotSpecifications
| MAST4 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 4930420O11Rik; AI642422; KIAA0303; MAST4; microtubule associated serine/threonine kinase family member 4; microtubule-associated serine/threonine-protein kinase 4; mKIAA0303; RGD1310139 | |
| Recombinant human MAST4 (immunogen range is 1431-1740). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| MAST4 |
| K56010_15H12 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O15021 | |
| 375449 | |
| IgG1 | |
| Protein A | |
| Antibody |