missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAST4 Monoclonal Antibody (K56010_15H12), Invitrogen™
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA556121
Description
Sequence of this protein is as follows: KSAEPPRSPL LKRVQSEEKL SPSYGSDKKH LCSRKHSLEV TQEEVQREQS QREAPLQSLD ENVCDVPPLS RARPVEQGCL KRPVSRKVGR QESVDDLDRD KLKAKVVVKK ADGFPEKQES HQKSHGPGSD LENFALFKLE EREKKVYPKA VERSSTFENK ASMQEAPPLG SLLKDALHKQ ASVRASEGAM SDGRVPAEHR QGGGDFRRAP APGTLQDGLC HSLDRGISGK GEGTEKSSQA KELLRCEKLD SKLANIDYLR KKMSLEDKED NLCPVLKPKM TAGSHECLPG NPVRPTGGQQ EPPPASESRA
The phosphorylation and dephosphorylation of proteins on serine and threonine residues is an essential means of regulating a broad range of cellular functions in eukaryotes, including cell division, homeostasis and apoptosis. A group of proteins that are intimately involved in this process are the serine/threonine(Ser/Thr) protein kinases. MAST4 (microtubule associated serine/threonine kinase family member 4), also known as KIAA0303, is a 2,626 amino acid protein that localizes to the cytoplasm and contains one protein kinase domain, as well as one PDZ domain and one AGC-kinase C-terminal domain. Expressed at high levels in most adult tissues, including testis, colon and small intestine, MAST4 uses magnesium as a cofactor to catalyze the ATP-dependent phosphorylation of target proteins. Multiple isoforms of MAST4 exist due to alternative splicing events.Chemical Identifiers
| MAST4 | |
| 1 mg/mL | |
| Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| 4930420O11Rik; AI642422; KIAA0303; MAST4; microtubule associated serine/threonine kinase family member 4; microtubule-associated serine/threonine-protein kinase 4; mKIAA0303; RGD1310139 | |
| Recombinant human MAST4 (immunogen range is 1431-1740). | |
| Primary | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| MAST4 |
| K56010_15H12 | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| O15021 | |
| 375449 | |
| IgG1 | |
| Protein A | |
| Antibody |
Specifications
| MAST4 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide | |
| O15021 | |
| MAST4 | |
| Recombinant human MAST4 (immunogen range is 1431-1740). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| K56010_15H12 | |
| Unconjugated | |
| MAST4 | |
| 4930420O11Rik; AI642422; KIAA0303; MAST4; microtubule associated serine/threonine kinase family member 4; microtubule-associated serine/threonine-protein kinase 4; mKIAA0303; RGD1310139 | |
| Mouse | |
| Protein A | |
| RUO | |
| 375449 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |