Learn More
MAPA™ UltraNeo™ 401 Polychloroprene and Natural Rubber Chemical Protection Gloves, Flock Lining
Gloves constructed of polychloroprene and natural rubber offer tactile sensitivity for light chemical protection.
$70.15 - $89.19
Specifications
Certifications/Compliance (Europe) Cat 3, CE 0334
Color Black
Latex Free No
Material Neoprene, Latex
Thickness (English) 22 mil
View More Specs
Products 5
Catalog Number Mfr. No. Size Price Quantity  
Catalog Number Mfr. No. Size Price Quantity  
11-392-34B
View Documents
MAPA™
401447
Medium
Dozen of 12 Pair for $72.88
Case of 6 Dozen for $437.28
Only null left
11-392-34C
View Documents
MAPA™
401448
Large
Dozen for $70.15
Case of 6 Dozen for $67.14
Only null left
11-392-34D
View Documents
MAPA™
401449
X-Large
Dozen for $89.19
Case of 6 Dozen for $535.14
Only null left
11-392-34E
View Documents
MAPA™
401440
2X-Large
Dozen of 12 Pair for $85.82
Case of 6 Dozen for $514.92
Only null left
19-050-602
View Documents
MAPA™
401441ZQK
3X-Large
Dozen of 12 Each for $72.53
Case of 6 Dozen for $435.18
Only null left
Description
Description
  • Multiple layer construction of neoprene outer layer and inner natural rubber
  • Embossed nonslip diamond grip for handling wet or greasy objects
  • Hand contoured for greater comfort
  • Cotton flock lining absorbs perspiration, maximizes easy on and off
  • Black
Specifications
Specifications
Cat 3, CE 0334
No
22 mil
EN 388:2110X
Type A
Bacteria, Fungi only
Embossed
Lined
12.5 in.
Black
Neoprene, Latex
0.55 mm
No
ALMNST
Straight
Flock
UltraNeo™ 401
32 cm
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'