missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Manganese Sulfate, Monohydrate, Powder, USP, 98-102%, Spectrum™ Chemical
M1114, 10034-96-5, MnSO4·H2O
Supplier: Spectrum Chemical Mfg Cor M111412KG
Description
Spectrum™ Chemical Manganese Sulfate, Monohydrate, Powder, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 10034-96-5 | |
| 12 kg | |
| MFCD00149159 | |
| O.[Mn++].[O-]S([O-])(=O)=O | |
| 169.01 | |
| USP | |
| Poly Pail |
| 100% | |
| H2MnO5S | |
| ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
| manganese(2+) hydrate sulfate | |
| 98 to 102% | |
| 10.0-13% |
Chemical Identifiers
| 10034-96-5 | |
| 169.01 | |
| ISPYRSDWRDQNSW-UHFFFAOYSA-L | |
| O.[Mn++].[O-]S([O-])(=O)=O |
| H2MnO5S | |
| MFCD00149159 | |
| manganese(2+) hydrate sulfate |