missing translation for 'onlineSavingsMsg'
Learn More

Manganese Sulfate, Monohydrate, Powder, USP, 98-102%, Spectrum™ Chemical

Catalog No. p-7157708
Change view
Click to view available options
Quantity:
500 g
2.5 kg
6 x 500 g
12 kg
4 x 2.5 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604208 500 g Poly Bottle
18604210 2.5 kg Poly Bottle
18604209 6 x 500 g Poly Bottle
18604211 4 x 2.5 kg Poly Bottle
18604212 12 kg Poly Pail
18604213 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18604208 Supplier Spectrum Chemical Mfg Cor Supplier No. M1114500GM

M1114, 10034-96-5, MnSO4·H2O

Spectrum™ Chemical Manganese Sulfate, Monohydrate, Powder, USP - All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 10034-96-5
Molecular Formula H2MnO5S
Molecular Weight (g/mol) 169.01
MDL Number MFCD00149159
InChI Key ISPYRSDWRDQNSW-UHFFFAOYSA-L
IUPAC Name manganese(2+) hydrate sulfate
SMILES O.[Mn++].[O-]S([O-])(=O)=O

Specifications

CAS Max % 100%
Quantity 500 g
Percent Purity 98 to 102%
Grade USP
Loss on Ignition 10.0-13%
Packaging Poly Bottle