missing translation for 'onlineSavingsMsg'
Learn More

Manganese Naphthenate (Mn ca. 6%), TCI America™

Catalog No. N034025G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N034025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N034025G Supplier TCI America Supplier No. N034025G
Only null left

Chemical Identifiers

CAS 1336-93-2
Molecular Formula C22H14MnO4
Molecular Weight (g/mol) 397.288
MDL Number MFCD00147533
InChI Key SGGOJYZMTYGPCH-UHFFFAOYSA-L
Synonym manganese ii naphthenate w/w in mineral spirits mn, manganese ii bis 2-naphthoate, manganese 2+ ;naphthalene-2-carboxylate, manganese 2+ bis 2-naphthoate
PubChem CID 25021872
IUPAC Name manganese(2+);naphthalene-2-carboxylate
SMILES C1=CC=C2C=C(C=CC2=C1)C(=O)[O-].C1=CC=C2C=C(C=CC2=C1)C(=O)[O-].[Mn+2]

Specifications

Color Black
Quantity 25 g
UN Number 1993
Physical Form Liquid
Chemical Name or Material Manganese Naphthenate (Mn ca. 6%)
EINECSNumber (8)-0630
RTECSNumber QK8985000
TSCA Yes