accessibility menu, dialog, popup

UserName

CAS RN 1336-93-2

IUPAC Name: manganese(2+) bis(naphthalene-2-carboxylate)
Synonyms: manganese(2+) bis(2-naphthoate)
Molecular Weight (g/mol): 397.29
Molecular Formula: C22H14MnO4
InChi Key: SGGOJYZMTYGPCH-UHFFFAOYSA-L
SMILES: [Mn++].[O-]C(=O)C1=CC2=CC=CC=C2C=C1.[O-]C(=O)C1=CC2=CC=CC=C2C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results