missing translation for 'onlineSavingsMsg'
Learn More

Maltitol Solution, NF, 50%, Spectrum™ Chemical

Catalog No. 18604158
Change view
Click to view available options
Quantity:
200 L
Packaging:
Steel Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18604158 200 L Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18604158 Supplier Spectrum Chemical Mfg Cor Supplier No. M1051200LTBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

M1051, 585-88-6

Spectrum™ Chemical Maltitol Solution, NF is used as a low-calorie sweetening agent in formulations. All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 585-88-6 , 7732-18-5
Molecular Formula C12H24O11
Molecular Weight (g/mol) 344.31
InChI Key VQHSOMBJVWLPSR-UHFFFAOYNA-N
IUPAC Name 4-{[3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy}hexane-1,2,3,5,6-pentol
SMILES OCC(O)C(O)C(OC1OC(CO)C(O)C(O)C1O)C(O)CO

Specifications

CAS Max % 50%
Ignition Residue 0.1%
Packaging Steel Drum
Quantity 200 L
Percent Purity 50%
Grade NF
Physical Form Liquid
Chemical Name or Material Maltitol Solution