Learn More
Thermo Scientific™ Male Luer-LOK Priming Syringes
Catalog No. 365KL531
Change view
Click to view available options
Volume (Metric):
1 mL
10 mL
Product Type:
Gas Tight GC Autosampler Syringe, Luer-Lok
GC Autosampler Syringe
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Product Type
365KL531 1 mL Gas Tight GC Autosampler Syringe, Luer-Lok
365N2721 10 mL GC Autosampler Syringe
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 365KL531 Supplier Thermo Scientific™ Supplier No. 365KL531
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
Thermo Scientific™ Male Luer-LOK Priming Syringes offer easy, accurate and reproducible manual injections.
  • Square tip to prevent damage to the injector
  • Wide range of volumes
  • Precision made from borosilicate glass and stainless steel
  • Robust design and easy-to-read markings
Specifications
Product Type Gas Tight GC Autosampler Syringe, Luer-Lok
Volume (Metric) 1 mL
For Use With (Equipment) Gas Chromatography Systems
Replacement Needle 365RNL22
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.