missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ MagniSort™ Streptavidin Positive Selection Beads

Designed for the magnetic separation of cells by positive selection. Desired cells are bound by biotinylated antibody and then streptavidin-coated magnetic beads. When placed in a magnetic field, desired cells are held in place and undesired cells can be separated by decanting.

$181.00 - $672.00

Specifications

Form Liquid
Ligand Streptavidin
Product Type Streptavidin Conjugate (Fluorescent)
Target Molecule Biotin
View More Specs

Alert:

Contains Sodium azide


Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501125939
View Documents
Invitrogen™
MSPB600374
200 Tests
Each for $672.00
Only null left
 
501125938
View Documents
Invitrogen™
MSPB600371
25 Tests
Each for $181.00
Only null left
 
Description

Description

  • Clone and concentration of biotinylated antibody for desired cell labeling, as well as concentration of MagniSort™ Streptavidin Positive Selection Beads must be determined empirically
  • Please refer to the protocol for details on how to optimize both the antibody and MagniSort™ Streptavidin Positive Selection Beads
  • The MagniSort™ Streptavidin Positive Selection Beads may also be used to deplete undesired cells using a single antibody
  • For depletion of cells using multiple antibodies, we recommend the MagniSort™ Streptavidin Negative Selection Beads (cat. MSNB-6002) and protocol
Specifications

Specifications

Liquid
Streptavidin
Streptavidin Conjugate (Fluorescent)
Biotin
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.