missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Trifluoromethanesulfonate 98.0+%, TCI America™

Supplier:  TCI America T13045G

 View more versions of this product

Catalog No. T13045G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Magnesium Trifluoromethanesulfonate
White
MFCD00042601
1759
BZQRBEVTLZHKEA-UHFFFAOYSA-L
magnesium;trifluoromethanesulfonate
2734125
≥98.0% (T)
60871-83-2
C2F6MgO6S2
5 g
magnesium trifluoromethanesulfonate, magnesium triflate, magnesium 2+ ditriflate, trifluoromethanesulfonic acid magnesium salt, magnesium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, magnesium salt, magnesiumtrifluoromethanesulfonate, magnesium trifiate, acmc-209mmv, ksc352a4p
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Mg+2]
322.431
322.43
Crystalline Powder

Chemical Identifiers

60871-83-2
322.431
BZQRBEVTLZHKEA-UHFFFAOYSA-L
2734125
C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Mg+2]
C2F6MgO6S2
MFCD00042601
magnesium trifluoromethanesulfonate, magnesium triflate, magnesium 2+ ditriflate, trifluoromethanesulfonic acid magnesium salt, magnesium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, magnesium salt, magnesiumtrifluoromethanesulfonate, magnesium trifiate, acmc-209mmv, ksc352a4p
magnesium;trifluoromethanesulfonate
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

TSCA : No

SDS
Documents

Documents

Promotions

Promotions