missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Trifluoromethanesulfonate 98.0+%, TCI America™

Catalog No. p-7136707
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T13045G 5 g
T130425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. T13045G Supplier TCI America Supplier No. T13045G
Only null left

Chemical Identifiers

CAS 60871-83-2
Molecular Formula C2F6MgO6S2
Molecular Weight (g/mol) 322.431
MDL Number MFCD00042601
InChI Key BZQRBEVTLZHKEA-UHFFFAOYSA-L
Synonym magnesium trifluoromethanesulfonate, magnesium triflate, magnesium 2+ ditriflate, trifluoromethanesulfonic acid magnesium salt, magnesium trifluoromethanesulphonate, methanesulfonic acid, trifluoro-, magnesium salt, magnesiumtrifluoromethanesulfonate, magnesium trifiate, acmc-209mmv, ksc352a4p
PubChem CID 2734125
IUPAC Name magnesium;trifluoromethanesulfonate
SMILES C(F)(F)(F)S(=O)(=O)[O-].C(F)(F)(F)S(=O)(=O)[O-].[Mg+2]

Specifications

Color White
Quantity 5 g
UN Number 1759
Formula Weight 322.43
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Magnesium Trifluoromethanesulfonate