accessibility menu, dialog, popup

UserName

FFFSOOOFFFSOOOMg

CAS RN 60871-83-2

IUPAC Name: magnesium(2+) ditrifluoromethanesulfonate
Synonyms: magnesium(2+) ditriflate
Molecular Weight (g/mol): 322.43
Molecular Formula: C2F6MgO6S2
InChi Key: BZQRBEVTLZHKEA-UHFFFAOYSA-L
SMILES: [Mg++].[O-]S(=O)(=O)C(F)(F)F.[O-]S(=O)(=O)C(F)(F)F
Quantity 
  • (1)
  • (2)
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (4)
  • (1)
  • (2)
  • (2)
  • (1)
  • (2)
  • (3)
  • (2)
  • (1)
  • (2)