missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Sulfate, For BOD, 2.25% (w/v), LabChem™

Catalog No. LC165001
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC165001 500 mL
LC165002 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC165001 Supplier LabChem Supplier No. LC165001
Only null left

Chemical Identifiers

CAS 10034-99-8 , 7732-18-5
Molecular Formula H14MgO11S
Molecular Weight (g/mol) 246.47
MDL Number MFCD00149785
InChI Key WRUGWIBCXHJTDG-UHFFFAOYSA-L
Synonym magnesium sulfate heptahydrate, mgso4.7h2o, magnesium sulfate 1:1 heptahydrate, magnesium sulphate heptahydrate, unii-sk47b8698t, magnesium sulfate usan:jan, sulfuric acid magnesium salt 1:1 , heptahydrate, magnesium sulfate heptahydrate mgso4.7h2o, sulfuric acid, magnesium salt 1:1 , heptahydrate, magnesium ii , sulfate, heptahydrate
PubChem CID 24843
ChEBI CHEBI:31795
IUPAC Name magnesium(2+) heptahydrate sulfate
SMILES O.O.O.O.O.O.O.[Mg++].[O-]S([O-])(=O)=O

Specifications

Color Colorless
CAS Max % 97.75, 2.25
Linear Formula MgSO4·7H2O
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 246.47
Percent Purity 2.25% (w/v)
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Sulfur compounds
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Magnesium Sulfate, For BOD
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature