missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E 530, light, 98.0-100.5% (calc. for dried substance), Honeywell Fluka™

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E 530, light, 98.0-100.5% (calc. for dried substance)

$458.43 - $716.88

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
6001966
View Documents
Honeywell-Fluka
13138500G
500 g
Each for $458.43
Only null left
 
6001965
View Documents
Honeywell-Fluka
131381KG
1 kg
Each for $716.88
Only null left
 
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

1309-48-4
40.30
CPLXHLVBOLITMK-UHFFFAOYSA-N
14792
O=[Mg]
MgO
MFCD00011109
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
oxomagnesium

Specifications

1309-48-4
MgO
MFCD00011109
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
O=[Mg]
40.30
40.3g/mol
puriss.
500 g
MgO
NONH for all modes of transport
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
14792
98 to 100.5% (calc. to the dried substance)
Magnesium Oxide, Meets analytical specification of Ph. Eur.BP, USP, FCC
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-171-9

RTECSNumber : OM3850000

SDS
missing translation for 'documents'

missing translation for 'documents'