missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, Puriss., meets analytical spec. of BP, FCC, Ph. Eur., USP, E 530, light, 98.0-100.5% (calc. for dried substance), Honeywell Fluka™

Catalog No. 6001966
Change view
Click to view available options
Quantity:
500 g
1 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
6001966 500 g
6001965 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001966 Supplier Honeywell Chemicals Supplier No. 13138500G
Only null left

Puriss., meets analytical specification of BP, FCC, Ph. Eur., USP, E 530, light, 98.0-100.5% (calc. for dried substance)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 1309-48-4
Molecular Formula MgO
Molecular Weight (g/mol) 40.30
MDL Number MFCD00011109
InChI Key CPLXHLVBOLITMK-UHFFFAOYSA-N
Synonym magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
PubChem CID 14792
IUPAC Name oxomagnesium
SMILES O=[Mg]

Specifications

Quantity 500 g
Linear Formula MgO
UN Number NONH for all modes of transport
Formula Weight 40.3g/mol
Percent Purity 98 to 100.5% (calc. to the dried substance)
Grade puriss.
Chemical Name or Material Magnesium Oxide, Meets analytical specification of Ph. Eur.BP, USP, FCC
EINECSNumber 215-171-9
RTECSNumber OM3850000