missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium oxide, 98%, extra pure, powder, particle size: 99% <150 μm (-100 mesh)
CAS: 1309-48-4 | MgO | 40.30 g/mol
$192.10 - $635.48
Chemical Identifiers
| CAS | 1309-48-4 |
|---|---|
| Molecular Formula | MgO |
| Molecular Weight (g/mol) | 40.30 |
| MDL Number | MFCD00011109 |
| InChI Key | CPLXHLVBOLITMK-UHFFFAOYSA-N |
| Synonym | magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag |
| PubChem CID | 14792 |
| IUPAC Name | oxomagnesium |
| SMILES | O=[Mg] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC263835000
|
Thermo Scientific Chemicals
263835000 |
500 g | Plastic bottle |
Each for $192.10
|
|
||||
|
AC263830025
|
Thermo Scientific Chemicals
263830025 |
2.5 kg | Plastic bottle |
Each for $635.48
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Chemical Identifiers
| 1309-48-4 | |
| 40.30 | |
| CPLXHLVBOLITMK-UHFFFAOYSA-N | |
| 14792 | |
| O=[Mg] |
| MgO | |
| MFCD00011109 | |
| magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
| oxomagnesium |
Specifications
| 3600.0°C | |
| 1309-48-4 | |
| Amorphous Powder | |
| 97.5% min. (Complexometry) | |
| MgO | |
| 01,633; 02,256; 07,221; 16,200 | |
| magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag | |
| CPLXHLVBOLITMK-UHFFFAOYSA-N | |
| oxomagnesium | |
| 14792 | |
| 98% | |
| 2% max. | |
| 3.58 | |
| Magnesium oxide, ca 200 Mesh |
| 2800.0°C | |
| White | |
| 500 g | |
| MgO | |
| MFCD00011109 | |
| 15, 5741 | |
| Solubility in water: 6.2mg/l (20°C),reacts generates heat and mg(oh)2. Other solubilities: soluble in diluted acids,insoluble in alcohol | |
| O=[Mg] | |
| 40.30 | |
| 40.29 | |
| Extra Pure | |
| Plastic bottle | |
| 3.5800g/mL |
Safety and Handling
EINECSNumber : 215-171-9
RUO – Research Use Only