missing translation for 'onlineSavingsMsg'
Learn More

Magnesium oxide, 98%, extra pure, powder, particle size: 99% <150 μm (-100 mesh)

CAS: 1309-48-4 | MgO | 40.30 g/mol

Supplier:  Thermo Scientific Chemicals 263830025

 View more versions of this product

Catalog No. AC263830025


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Specifications

Specifications

3600.0°C
2800.0°C
1309-48-4
Amorphous Powder
97.5% min. (Complexometry)
MgO
01,633; 02,256; 07,221; 16,200
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
CPLXHLVBOLITMK-UHFFFAOYSA-N
oxomagnesium
14792
98%
2% max.
3.58
Magnesium oxide
ca 200 Mesh
White
2.5 kg
MgO
MFCD00011109
15, 5741
Solubility in water: 6.2mg/l (20°C),reacts generates heat and mg(oh)2. Other solubilities: soluble in diluted acids,insoluble in alcohol
O=[Mg]
40.30
40.29
Extra Pure
Plastic bottle
3.5800g/mL

Chemical Identifiers

1309-48-4
40.30
CPLXHLVBOLITMK-UHFFFAOYSA-N
14792
O=[Mg]
MgO
MFCD00011109
magnesium oxide, magnesia, seawater magnesia, causmag, granmag, maglite, periclase, seasorb, animag
oxomagnesium
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : 215-171-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only