missing translation for 'onlineSavingsMsg'
Learn More

Magnesium EDTA, For Hardness, 0.02% (w/v), LabChem™

Catalog No. p-5388418
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
LC164501 500 mL
LC164502 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. LC164501 Supplier LabChem Supplier No. LC164501

Chemical Identifiers

CAS 14402-88-1 , 7732-18-5
Molecular Formula C10H12MgN2Na2O8
Molecular Weight (g/mol) 358.50
MDL Number MFCD00078217
InChI Key AWNVVAMWLMUZOZ-UHFFFAOYSA-J
Synonym unii-ndt563s5vz, edta magnesium disodium, magnesium disodium edta, ndt563s5vz, magnesium disodium ethylenediaminetetraacetate, edta magnesium disodium salt hydrate, ethylenediaminetetraacetic acid magnesium disodium salt, ethylenediaminetetraacetic acid disodium magnesium salt, ethylenediaminetetraacetic acid disodium magnesium salt hydrate, magnesium sodium ethylenediaminetetraacetate
PubChem CID 161064
IUPAC Name magnesium(2+) disodium 2-({2-[bis(carboxylatomethyl)amino]ethyl}(carboxylatomethyl)amino)acetate
SMILES [Na+].[Na+].[Mg++].[O-]C(=O)CN(CCN(CC([O-])=O)CC([O-])=O)CC([O-])=O

Specifications

Color Colorless
CAS Max % 99.98, 0.02
Linear Formula C10H12N2O8MgNa2
Density 1g/mL
Solubility Information Soluble in water
Formula Weight 358.5
Percent Purity 0.02% (w/v)
Identification Passes Test
Packaging Poly Bottle
Specific Gravity 1g/mL
Decomposition Information Nitrogen oxides; Carbon monoxide; Carbon dioxide
Quantity 500 mL
Physical Form Liquid
Chemical Name or Material Magnesium EDTA, For Hardness
Show More Show Less
Hazard Statement GHS H Statement
Solution is not hazardous.
Precautionary Statement GHS P Statement
If in contact with skin or eyes, rinse thoroughly with water for 15-20 minutes.
If swallowed, get medical attention.
Recommended Storage Room Temperature