missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride, Hexahydrate, Crystal, ACS, 99-102%, Spectrum™ Chemical
$190.09 - $6571.59
Chemical Identifiers
| CAS | 7791-18-6 |
|---|---|
| Molecular Formula | Cl2H12MgO6 |
| Molecular Weight (g/mol) | 203.30 |
| MDL Number | MFCD00149781 |
| InChI Key | DHRRIBDTHFBPNG-UHFFFAOYSA-L |
| IUPAC Name | magnesium(2+) hexahydrate dichloride |
| SMILES | O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18611415
|
Spectrum Chemical Mfg Cor
M103525GM |
25 g | Poly Bottle |
Each for $190.09
|
|
||||
|
18611416
|
Spectrum Chemical Mfg Cor
M1035125GM |
125 g | Poly Bottle |
Each for $300.15
|
|
||||
|
18611417
|
Spectrum Chemical Mfg Cor
M1035500GM |
500 g | Poly Bottle |
Each for $337.65
|
|
||||
|
18611418
|
Spectrum Chemical Mfg Cor
M103525KG |
2.5 kg | Poly Bottle |
Each for $1,482.68
|
|
||||
|
18611419
|
Spectrum Chemical Mfg Cor
M103512KG |
12 kg | Poly Pail |
Each for $3,069.24
|
|
||||
|
18611420
|
Spectrum Chemical Mfg Cor
M103550KGBL |
50 kg | Fiber Drum |
Each for $6,571.59
|
|
||||
Chemical Identifiers
| 7791-18-6 | |
| 203.30 | |
| DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
| O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
| Cl2H12MgO6 | |
| MFCD00149781 | |
| magnesium(2+) hexahydrate dichloride |
Specifications
| 7791-18-6 | |
| 25 g | |
| MFCD00149781 | |
| O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] | |
| 203.30 | |
| ACS |
| 1 | |
| Cl2H12MgO6 | |
| DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
| magnesium(2+) hexahydrate dichloride | |
| 99 to 102% | |
| Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.