missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride, Hexahydrate, Crystal, ACS, 99-102%, Spectrum™ Chemical

Catalog No. 18611415
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611415 25 g Poly Bottle
18611416 125 g Poly Bottle
18611417 500 g Poly Bottle
18611418 2.5 kg Poly Bottle
18611419 12 kg Poly Pail
18611420 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18611415 Supplier Spectrum Chemical Mfg Cor Supplier No. M103525GM
Only null left

Chemical Identifiers

CAS 7791-18-6
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

CAS Max % 1
Quantity 25 g
Percent Purity 99 to 102%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.