missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Chloride, Hexahydrate, Crystal, ACS, 99-102%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor M1035500GM
Specifications
| 7791-18-6 | |
| 500 g | |
| MFCD00149781 | |
| O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] | |
| 203.30 | |
| ACS |
| 1 | |
| Cl2H12MgO6 | |
| DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
| magnesium(2+) hexahydrate dichloride | |
| 99 to 102% | |
| Poly Bottle |
Chemical Identifiers
| 7791-18-6 | |
| 203.30 | |
| DHRRIBDTHFBPNG-UHFFFAOYSA-L | |
| O.O.O.O.O.O.[Mg++].[Cl-].[Cl-] |
| Cl2H12MgO6 | |
| MFCD00149781 | |
| magnesium(2+) hexahydrate dichloride |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.