missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Chloride, 51% (w/v), Ricca Chemical

Catalog No. 447016
Change view
Click to view available options
Quantity:
500 mL
1 L
Packaging:
Natural Poly Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
447016 500 mL Natural Poly Bottle
447032 1 L Natural Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 447016 Supplier Ricca Chemical Company Supplier No. 447016
Only null left

Chemical Identifiers

CAS 7791-18-6 , 7732-18-5
Molecular Formula Cl2H12MgO6
Molecular Weight (g/mol) 203.30
MDL Number MFCD00149781
InChI Key DHRRIBDTHFBPNG-UHFFFAOYSA-L
IUPAC Name magnesium(2+) hexahydrate dichloride
SMILES O.O.O.O.O.O.[Mg++].[Cl-].[Cl-]

Specifications

Color Colorless
Percent Purity 51%
Grade Laboratory
Packaging Natural Poly Bottle
Solvent or Matrix Aqueous Solution
pH Neutral
Quantity 500 mL
Physical Form Liquid
Concentration or Composition Notes 51% (w/v)
Chemical Name or Material Magnesium Chloride Solution