missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Acetate, Tetrahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical
$299.53 - $13063.67
Chemical Identifiers
| CAS | 16674-78-5 |
|---|---|
| Molecular Formula | C4H14MgO8 |
| Molecular Weight (g/mol) | 214.45 |
| MDL Number | MFCD00149214 |
| InChI Key | XKPKPGCRSHFTKM-UHFFFAOYSA-L |
| IUPAC Name | magnesium(2+) diacetate tetrahydrate |
| SMILES | O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
18611399
|
Spectrum Chemical Mfg Cor
M1020125GM |
125 g | Poly Bottle |
Each for $299.53
|
|
||||
|
18611400
|
Spectrum Chemical Mfg Cor
M1020500GM |
500 g | Poly Bottle |
Each for $790.53
|
|
||||
|
18611401
|
Spectrum Chemical Mfg Cor
M102025KG |
2.5 kg | Poly Bottle |
Each for $2,491.87
|
|
||||
|
18611402
|
Spectrum Chemical Mfg Cor
M102012KG |
12 kg | Poly Pail |
Each for $5,914.53
|
|
||||
|
18611403
|
Spectrum Chemical Mfg Cor
M102045KGBL |
45 kg | Fiber Drum |
N/A
|
N/A | ||||
| Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance. | |||||||||
Chemical Identifiers
| 16674-78-5 | |
| 214.45 | |
| XKPKPGCRSHFTKM-UHFFFAOYSA-L | |
| O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O |
| C4H14MgO8 | |
| MFCD00149214 | |
| magnesium(2+) diacetate tetrahydrate |
Specifications
| 16674-78-5 | |
| 125 g | |
| MFCD00149214 | |
| O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O | |
| 214.45 | |
| ACS |
| 1 | |
| C4H14MgO8 | |
| XKPKPGCRSHFTKM-UHFFFAOYSA-L | |
| magnesium(2+) diacetate tetrahydrate | |
| 98 to 102% | |
| Poly Bottle |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.