missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Magnesium Acetate, Tetrahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical
Supplier: Spectrum Chemical Mfg Cor M102025KG
Specifications
| 16674-78-5 | |
| 2.5 kg | |
| MFCD00149214 | |
| O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O | |
| 214.45 | |
| ACS |
| 1 | |
| C4H14MgO8 | |
| XKPKPGCRSHFTKM-UHFFFAOYSA-L | |
| magnesium(2+) diacetate tetrahydrate | |
| 98 to 102% | |
| Poly Bottle |
Chemical Identifiers
| 16674-78-5 | |
| 214.45 | |
| XKPKPGCRSHFTKM-UHFFFAOYSA-L | |
| O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O |
| C4H14MgO8 | |
| MFCD00149214 | |
| magnesium(2+) diacetate tetrahydrate |
CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.