missing translation for 'onlineSavingsMsg'
Learn More

Magnesium Acetate, Tetrahydrate, Crystal, ACS, 98-102%, Spectrum™ Chemical

Catalog No. 18611403
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
45 kg
Packaging:
Amber Glass Bottle
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18611403 45 kg Poly Bottle
18611399 125 g Amber Glass Bottle
18611400 500 g Poly Bottle
18611401 2.5 kg Poly Bottle
18611402 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18611403 Supplier Spectrum Chemical Mfg Cor Supplier No. M102045KGBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 16674-78-5
Molecular Formula C4H14MgO8
Molecular Weight (g/mol) 214.45
MDL Number MFCD00149214
InChI Key XKPKPGCRSHFTKM-UHFFFAOYSA-L
IUPAC Name magnesium(2+) diacetate tetrahydrate
SMILES O.O.O.O.[Mg++].CC([O-])=O.CC([O-])=O

Specifications

Physical Form Crystalline Powder or Needles
CAS Max % 1
Quantity 45 kg
Packaging Poly Bottle
Percent Purity 98 to 102%
Grade ACS/Reagent
Chemical Name or Material Magnesium Acetate, Tetrahydrate, Crystal

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.